Merck
  • The α₁-, but not α₂-, adrenoceptor in the nucleus accumbens plays an inhibitory role upon the accumbal noradrenaline and dopamine efflux of freely moving rats.
The α₁-, but not α₂-, adrenoceptor in the nucleus accumbens plays an inhibitory role upon the accumbal noradrenaline and dopamine efflux of freely moving rats.
European journal of pharmacology (2012-05-24)
Tadashi Saigusa, Yuri Aono, Takuya Uchida, Koji Takada, Michel M M Verheij, Noriaki Koshikawa, Alexander R Cools
ABSTRACT
In vivo microdialysis was used to analyse the role of the α(1)- and α(2)-adrenoceptor subtypes in the regulation of noradrenaline and dopamine efflux in the nucleus accumbens of freely moving rats. Intra-accumbal infusion of α(1)-adrenoceptor agonist methoxamine (24pmol) failed to alter the noradrenaline efflux, but decreased the dopamine efflux. The intra-accumbal infusion of α(1)-adrenoceptor antagonist prazosin (6, 600 and 6000pmol) produced a dose-related increase and decrease of the noradrenaline and dopamine efflux, respectively. An ineffective dose of prazosin (6pmol) counteracted the methoxamine (24pmol)-induced decrease of dopamine efflux. The prazosin (6000pmol)-induced increase of noradrenaline efflux, but not the decrease of dopamine efflux, was suppressed by the co-administration of an ineffective dose of methoxamine (0.024pmol). Neither the α(2)-adrenoceptor agonist clonidine (300pmol) and UK 14,304 (300pmol) nor the α(2)-adrenoceptor antagonist RX 821002 (0.6, 3, 600 and 6000pmol) significantly affected the accumbal noradrenaline and dopamine efflux. The doses mentioned are the total amount of drug over the 60-min infusion period. The present results show that (1) accumbal α(1)-adrenoceptors which are presynaptically located on noradrenergic nerve terminals inhibit the accumbal noradrenaline efflux, increasing thereby the accumbal dopamine efflux, (2) accumbal α(1)-adrenoceptors which are postsynaptically located on dopaminergic nerve terminals inhibit the accumbal dopamine efflux, and (3) accumbal α(2)-adrenoceptors play no major role in the regulation of accumbal efflux of noradrenaline and dopamine.
Sigma-Aldrich
Idazoxan hydrochloride
Sigma-Aldrich
Methoxamine hydrochloride
Sigma-Aldrich
UK 14,304
Sigma-Aldrich
RX 821002 hydrochloride, solid, ≥98% (HPLC)