Merck
  • O-(2,3,4,5,6-pentafluorophenyl)methylhydroxylamine hydrochloride: a versatile reagent for the determination of carbonyl-containing compounds.
O-(2,3,4,5,6-pentafluorophenyl)methylhydroxylamine hydrochloride: a versatile reagent for the determination of carbonyl-containing compounds.
Journal of chromatography (1992-12-25)
D A Cancilla, S S Que Hee
ABSTRACT
A review on the use of O-(2,3,4,5,6-pentafluorophenyl)methylhydroxylamine hydrochloride (PFBHA) for the determination of carbonyl-containing compounds is presented. PFBHA has been used in the determination of such diverse compounds as thromboxane B2, prostaglandins, amygdalin and a variety of other aldehydes, ketones and acids. PFBHA has been used for the determination of these compounds found in water, blood, urine, air and even clothing. The review covers literature referenced in Chemical Abstracts from 1975, when PFBHA was first synthesized, through March 1992.
Sigma-Aldrich
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, ≥98%
Supelco
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, derivatization grade (GC derivatization), LiChropur, ≥99.0% (AT)