Merck
Protecting group-free total synthesis of (-)-lannotinidine B.
Journal of the American Chemical Society (2012-07-18)
Hui Ming Ge, Lan-De Zhang, Ren Xiang Tan, Zhu-Jun Yao
ABSTRACT
The first total synthesis of (-)-lannotinidine B, a unique tetracyclic constitutent of Lycopodium annotinum, has been accomplished in 10 steps with 23% overall yield. The completed short and efficient synthesis is characterized with three highly chemo- and/or stereoselective reductive-amination steps to furnish the desired trans-fused 6/6 bicycle and the aza seven-membered ring system, and a direct intramolecular acyloin condensation to deliver the cyclopentanone moiety, as well as successful application of a protecting group-free strategy and an optimal redox order.
Sigma-Aldrich
Cyclopentanone, ReagentPlus®, ≥99%
Supelco
Cyclopentanone, analytical standard
Sigma-Aldrich
Cyclopentanone, ≥99%, FG