Merck
  • Iron-lactoferrin complex reduces iron-catalyzed off-flavor formation in powdered milk with added fish oil.
Iron-lactoferrin complex reduces iron-catalyzed off-flavor formation in powdered milk with added fish oil.
Journal of food science (2012-08-07)
Hiroshi M Ueno, Makoto Shiota, Noriko Ueda, Tomoyuki Isogai, Toshiya Kobayashi
ABSTRACT
The iron-lactoferrin complex (FeLf) is useful for dietary iron supplementation. However, the effects of FeLf on iron-catalyzed off-flavors in lipid-containing food products have not been reported. In this study, we investigated the effects of FeLf on off-flavors development during the production and storage of powdered milk with added fish oil. Powdered milk samples were formulated with FeLf or iron (II) sulfate, then stored at 37 °C for 5 mo. A sensory evaluation revealed that FeLf delayed the development of oxidized flavor and reduced metallic taste in the powdered milk compared with iron (II) sulfate. Headspace gas chromatography-mass spectrometry analysis showed that oxidized volatile compounds, such as pentanal, hexanal, heptanal, octanal, 1-penten-3-one, (Z)-4-heptenal, (E, E)-2,4-heptadienal, and (E)-2-octenal, were less developed in the powdered milk containing FeLf than in that containing iron (II) sulfate. Iron and lactoferrin (Lf) were retained in the high-molecular-weight (>10000 Da) fraction of the reconstituted FeLf-containing powdered milk after its manufacture and storage, whereas the antigenicity of Lf was lost after ultrahigh-temperature processing at 120 °C for 5 s. These results suggest that FeLf reduces the iron-catalyzed off-flavors that develop during the production and storage of powdered milk. The stable iron-holding property of FeLf contributes to the inhibition of oxidized and metallic volatile formation, although the loss of Lf antigenicity did not affect the stability of FeLf and the iron-catalyzed formation of off-flavors in the powdered milk. Consequently, FeLf is a suitable candidate for the simultaneous supplementation of a single food with iron and fish oil. The supplementation of food products with iron and fish oil is a useful approach to redressing their inadequate intake in many populations. The iron-lactoferrin complex can protect food products against the off-flavors caused by iron-catalyzed lipid oxidation. Our results show that the iron-lactoferrin complex is useful for the simultaneous fortification of foods and nutraceuticals with iron and fish oil because this complex also reduces the formation of iron-catalyzed off-flavors in powdered milk enriched with fish oil.
Sigma-Aldrich
trans-2-Octenal, technical grade, 94%
Sigma-Aldrich
trans-2-Octenal, ≥95%, stabilized, FG
Sigma-Aldrich
Hexanal, ≥97%, FCC, stabilized, FG
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Valeraldehyde, 97%
Sigma-Aldrich
Heptaldehyde, 95%
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Sigma-Aldrich
Valeraldehyde, ≥97%, FG