Merck
  • Stimulation of healing within a rabbit calvarial defect by a PCL/PLGA scaffold blended with TCP using solid freeform fabrication technology.
Stimulation of healing within a rabbit calvarial defect by a PCL/PLGA scaffold blended with TCP using solid freeform fabrication technology.
Journal of materials science. Materials in medicine (2012-09-11)
Jin-Hyung Shim, Tae-Sung Moon, Mi-Jung Yun, Young-Chan Jeon, Chang-Mo Jeong, Dong-Woo Cho, Jung-Bo Huh
ABSTRACT
The purpose of this study was to investigate the healing capacity within an 8-mm rabbit calvarial defect using a polycaprolactone (PCL)/poly(lactic-co-glycolic acid) (PLGA) scaffold blended with tri-calcium phosphate (TCP) that was constructed using solid freeform fabrication (SFF) technology. The PCL/PLGA/TCP scaffold showed a 37 % higher compressive strength and rougher surface than the PCL/PLGA scaffold. In animal experiments, new bone formation was analyzed using microcomputed tomography (micro-CT) and histological and histometric analyses. The PCL/PLGA/TCP groups had significantly greater neo-tissue areas as compared with the control groups at 4 and 8 weeks (P < 0.05). The PCL/PLGA/TCP group had significantly greater bone density as compared with the control and PCL/PLGA groups at 4 and 8 weeks (P < 0.005). The results of this study suggest that the PCL/PLGA/TCP scaffold fabricated using SFF technology is useful for recovering and enhancing new bone formation in bony defects in rabbits.
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Sigma-Aldrich
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)