Merck
[Halitosis in certain somatic diseases].
Stomatologiia (2012-09-13)
D N Bakhmutov, I G Fedorov, O I Kharchenko, O O Ianushevich
ABSTRACT
The structure of exhaled air at various somatopathies is evaluated in the study. Presence and concentration of hydrogen sulfide, dimethilsulfide, putrescine, cadaverine in patients with otolaryngological, endocrine pathology, diseases of gastrointestinal tract and oral diseases. The obtained data is of great importance for halitosis causes diagnostics.
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Sodium thiomethoxide, 95%
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Supelco
Dimethyl sulfide, analytical standard
Sigma-Aldrich
Sodium methanethiolate, technical, ≥90% (RT)