Merck
  • Radiation-induced impacts on the degradation of 2,4-D and the microbial population in soil microcosms.
Radiation-induced impacts on the degradation of 2,4-D and the microbial population in soil microcosms.
Journal of environmental radioactivity (2012-09-15)
Bastian Niedrée, Harry Vereecken, Peter Burauel
ABSTRACT
In a soil microcosm experiment, the influence of low-level (137)Cs and (90)Sr contamination on the degradation of (14)C-ring-labeled 2,4-dichlorophenoxyacetic acid (2,4-D) was studied. Two differently treated soils (one native soil and one soil sterilized and reinoculated with a biotic soil aliquot) were artificially contaminated with various concentrations of (137)Cs and (90)Sr as nitrate salts. The cumulative doses increased up to 4 Gy for 30 days of incubation in soil microcosms. Changes in microbial community structure were observed with help of the denaturing gradient gel electrophoresis (DGGE). A radiation-induced impact appeared only in the microcosms treated with 30 times the maximum contamination appearing in the exclusion zone around reactor 4 in Chernobyl. In contrast to the less contaminated soils, the mineralization of 2,4-D was delayed for 4 days before it recovered. Slight shifts in the microbial communities could be traced to radiation effects. However, other parameters had a major impact on mineralization and community structure. Thus the sterilization and reinoculation and, of course, application of the 2,4-D were predominantly reflected in the (14)CO(2) emissions and the DGGE gel patterns.
Product Description
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline