Merck
  • Restoration of dopamine release deficits during object recognition memory acquisition attenuates cognitive impairment in a triple transgenic mice model of Alzheimer's disease.
Restoration of dopamine release deficits during object recognition memory acquisition attenuates cognitive impairment in a triple transgenic mice model of Alzheimer's disease.
Learning & memory (Cold Spring Harbor, N.Y.) (2012-09-18)
Kioko Guzmán-Ramos, Perla Moreno-Castilla, Monica Castro-Cruz, James L McGaugh, Hilda Martínez-Coria, Frank M LaFerla, Federico Bermúdez-Rattoni
ABSTRACT
Previous findings indicate that the acquisition and consolidation of recognition memory involves dopaminergic activity. Although dopamine deregulation has been observed in Alzheimer's disease (AD) patients, the dysfunction of this neurotransmitter has not been investigated in animal models of AD. The aim of this study was to assess, by in vivo microdialysis, cortical and hippocampal dopamine, norepinephrine, and glutamate release during the acquisition of object recognition memory (ORM) in 5- and 10-mo-old triple-transgenic Alzheimer's disease mice (3xTg-AD) and to relate the extracellular changes to 24-h memory performance. Five- and 10-mo-old wild-type mice and 5-mo-old 3xTg-AD showed significant cortical but not hippocampal dopamine increase during object exploration. On a 24-h ORM test, these three groups displayed significant ORM. In contrast, 10-mo-old 3xTg-AD mice showed impaired dopamine release in the insular cortex during ORM acquisition, as well as significant impairment in ORM. In addition, cortical administration of a dopamine reuptake blocker produced an increase of dopamine levels in the 10-mo-old 3xTg-AD mice and attenuated the memory impairment. These data suggest that activation of the dopaminergic system in the insular cortex is involved in object recognition memory, and that dysfunction of this system contributes to the age-related decline in cognitive functioning of the 3xTg-AD mice.
Sigma-Aldrich
Nomifensine maleate salt