Merck
  • Experimental and theoretical investigation of 1J(CC) and (n)J(CC) coupling constants in strychnine.
Experimental and theoretical investigation of 1J(CC) and (n)J(CC) coupling constants in strychnine.
Organic letters (2012-09-22)
R Thomas Williamson, Alexei V Buevich, Gary E Martin
ABSTRACT
A relatively unexplored and unexploited means of establishing molecular structure, stereochemistry, and probing vicinal bond angles is through the use of long-range (13)C-(13)C coupling constants. The measurement of these multifunctional, diagnostic (3)J(CC) couplings has not been reported on sample amounts viable for the practicing organic chemist. A generalized protocol for the measurement of (1)J(CC) and (3)J(CC) couplings using a 4.6 mg sample of strychnine as a model compound is described, and the utility of DFT calculations for the prediction of these useful molecular descriptors and the congruence of the calculated and experimental data is demonstrated.
Sigma-Aldrich
Strychnine