Merck
  • Serum glutamate levels correlate with Gleason score and glutamate blockade decreases proliferation, migration, and invasion and induces apoptosis in prostate cancer cells.
Serum glutamate levels correlate with Gleason score and glutamate blockade decreases proliferation, migration, and invasion and induces apoptosis in prostate cancer cells.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-10-18)
Shahriar Koochekpour, Sunipa Majumdar, Gissou Azabdaftari, Kristopher Attwood, Ray Scioneaux, Dhatchayini Subramani, Charles Manhardt, Giovanni D Lorusso, Stacey S Willard, Hillary Thompson, Mojgan Shourideh, Katayoon Rezaei, Oliver Sartor, James L Mohler, Robert L Vessella
ABSTRACT
During glutaminolysis, glutamine is catabolized to glutamate and incorporated into citric acid cycle and lipogenesis. Serum glutamate levels were measured in patients with primary prostate cancer or metastatic castrate-resistant prostate cancer (mCRPCa) to establish clinical relevance. The effect of glutamate deprivation or blockade by metabotropic glutamate receptor 1 (GRM1) antagonists was investigated on prostate cancer cells' growth, migration, and invasion to establish biologic relevance. Serum glutamate levels were measured in normal men (n = 60) and patients with primary prostate cancer (n = 197) or mCRPCa (n = 109). GRM1 expression in prostatic tissues was examined using immunohistochemistry (IHC). Cell growth, migration, and invasion were determined using cell cytotoxicity and modified Boyden chamber assays, respectively. Apoptosis was detected using immunoblotting against cleaved caspases, PARP, and γ-H2AX. Univariate and multivariate analyses showed significantly higher serum glutamate levels in Gleason score ≥ 8 than in the Gleason score ≤ 7 and in African Americans than in the Caucasian Americans. African Americans with mCRPCa had significantly higher serum glutamate levels than those with primary prostate cancer or benign prostate. However, in Caucasian Americans, serum glutamate levels were similar in normal research subjects and patients with mCRPC. IHC showed weak or no expression of GRM1 in luminal acinar epithelial cells of normal or hyperplastic glands but high expression in primary or metastatic prostate cancer tissues. Glutamate deprivation or blockade decreased prostate cancer cells' proliferation, migration, and invasion and led to apoptotic cell death. Glutamate expression is mechanistically associated with and may provide a biomarker of prostate cancer aggressiveness.
Sigma-Aldrich
Riluzole, solid