Merck
  • Analysis of new potential anticonvulsant compounds in mice brain tissue by SPE/HPLC/DAD.
Analysis of new potential anticonvulsant compounds in mice brain tissue by SPE/HPLC/DAD.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-11-17)
Jolanta Flieger, Magdalena Pizoń, Tomasz Plech, Jarogniew J Luszczki
ABSTRACT
This paper describes a novel reversed-phase high performance liquid chromatography (RP-HPLC) with photo-diode array detection (DAD) method for the determination of three new derivatives of 4-alkyl-5-(3-chlorophenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione with different antiepileptic activity in the brains of mice treated with the doses of 300mgkg(-1) of body weight. Samples were prepared by solid-phase extraction (SPE) method using BAKERBOND™ spe Octadecyl (C(18)) and analyzed by the use of an isocratic elution mode over an Zorbax Extend-C18 column (150mm×4.6mm I.D., 5-μm, Agilent Technologies). The mobile phase consisted of 80% methanol (for compound TP-315) and 85% acetonitrile (for compound TP-321) for 80% 2-propanol (for TP-323) at a flow rate of 1.0mLmin(-1) and 0.5mLmin(-1) in the last case. Gradient elution mode was also proposed for all examined analytes in mixture with common antiepileptic drugs: carbamazepine, phenobarbital and phenytoin in view of possible synergistic activity. Photodiode-array investigations of the peaks after degradation studies indicate the stability of the compounds under conditions proposed for sample preparation procedure. Linear coefficients of correlation (r(2)) were >0.995 for all analytes. The proposed strategy gives extraction yields higher than 95% with the intra- and inter-day relative standard deviation lower than 3% and 5%, respectively. This method was applied to the analysis of brain tissue of mice treated with investigated compounds. Obtained results enable to explain the differences in their pharmacological activity.
Sigma-Aldrich
1,2,4-Triazole, 98%