Merck
  • Crystal structures of HIV-1 reverse transcriptase with picomolar inhibitors reveal key interactions for drug design.
Crystal structures of HIV-1 reverse transcriptase with picomolar inhibitors reveal key interactions for drug design.
Journal of the American Chemical Society (2012-11-21)
Kathleen M Frey, Mariela Bollini, Andrea C Mislak, José A Cisneros, Ricardo Gallardo-Macias, William L Jorgensen, Karen S Anderson
ABSTRACT
X-ray crystal structures at 2.9 Å resolution are reported for two complexes of catechol diethers with HIV-1 reverse transcriptase. The results help elucidate the structural origins of the extreme antiviral activity of the compounds. The possibility of halogen bonding between the inhibitors and Pro95 is addressed. Structural analysis reveals key interactions with conserved residues P95 and W229 of importance for design of inhibitors with high potency and favorable resistance profiles.
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Bromine, reagent grade
Sigma-Aldrich
Bromine, ACS reagent, ≥99.5%