Merck
  • Fabrication and evaluation of pH-modulated solid dispersion for telmisartan by spray-drying technique.
Fabrication and evaluation of pH-modulated solid dispersion for telmisartan by spray-drying technique.
International journal of pharmaceutics (2012-11-24)
Nirmal Marasini, Tuan Hiep Tran, Bijay Kumar Poudel, Hyuk Jun Cho, Young Keun Choi, Sang-Cheol Chi, Han-Gon Choi, Chul Soon Yong, Jong Oh Kim
ABSTRACT
The present study was undertaken to overcome the problems associated with solubility, dissolution and oral bioavailability of a poorly water-soluble ionizable drug, telmisartan (TMS). For these purposes, a solubility test was carried to select the appropriate formulation composition from various carriers and alkalizers. Solid dispersions (SDs) of TMS were prepared at different drug-to-carrier ratios by the spray-drying technique, and were characterized by dissolution and aqueous solubility studies. The optimum formulation was investigated by dissolution studies at different pH and water media and its solid state characterisations were performed by scanning electron microscopy (SEM), differential scanning calorimetry (DSC) and X-ray diffraction (XRD) studies. In solubility and dissolution tests, all TMS-loaded pH-modulated SDs (pH(M)-SDs) exhibited marked improvement in the dissolution behavior when compared with crystalline TMS powder. The optimum formulation of pH(M)-SD consisted of TMS/PVP (polyvinylpyrrolidone) K30/Na(2)CO(3) at a weight ratio of 2/0.5/3 and showed significant improvement in the aqueous solubility and dissolution rate by approximately 40,000- and 3-fold, respectively, compared to TMS powder. Solid-state characterization revealed the changed in crystallinity of TMS into amorphous state. Furthermore, area under the drug concentration time-curve (AUC) of TMS from the pH(M)-SD increased by 13.4- and 2.1-fold, compared with TMS powder and commercial product, respectively. According to these observations, taken together with dissolution and pharmacokinetic behaviors, pH-modulated SD in the presence of an alkalizer for a poorly water-soluble ionizable drug, TMS, appeared to be efficacious for enhancing its bioavailability.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C