Merck
  • Response of bacterial isolates from Antarctic shallow sediments towards heavy metals, antibiotics and polychlorinated biphenyls.
Response of bacterial isolates from Antarctic shallow sediments towards heavy metals, antibiotics and polychlorinated biphenyls.
Ecotoxicology (London, England) (2012-11-28)
Angelina Lo Giudice, Patrizia Casella, Vivia Bruni, Luigi Michaud
ABSTRACT
The response of bacterial isolates from Antarctic sediments to polychlorinated biphenyls (Aroclor 1242 mixture), heavy metal salts (cadmium, copper, mercury and zinc) and antibiotics (ampicillin, chloramphenicol, kanamycin and streptomycin) was investigated. Overall, the ability to growth in the presence of Aroclor 1242 as a sole carbon source was observed for 22 isolates that mainly belonged to Psychrobacter spp. Tolerance to the heavy metals assayed in this study was in the order of Cd > Cu > Zn > Hg and appeared to be strictly related to the metal concentrations, as determined during previous chemical surveys in the same area. With regards to antibiotic assays, the response of the isolates to the tested antibiotics ranged from complete resistance to total susceptibility. In particular, resistances to ampicillin and chloramphenicol were very pronounced in the majority of isolates. Our isolates differently responded to the presence of toxic compounds primarily based on their phylogenetic affiliation and secondarily at strain level. Moreover, the high incidence of resistance either to metal or antibiotics, in addition to the capability to grow on PCBs, confirm that bacteria are able to cope and/or adapt to the occurrence pollutants even in low human-impacted environments.
Product Description
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Cadmium chloride
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Supelco
Aroclor 1242 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Aroclor 1242 solution, certified reference material, 500 mg/kg in transformer oil
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture