Merck
  • Selenium (Se) seed priming induced growth and biochemical changes in wheat under water deficit conditions.
Selenium (Se) seed priming induced growth and biochemical changes in wheat under water deficit conditions.
Biological trace element research (2012-12-01)
Fahim Nawaz, M Yasin Ashraf, Rashid Ahmad, Ejaz Ahmad Waraich
ABSTRACT
Insufficient stand establishment at early growth stages in wheat (Triticum aestivum L.) due to drought stress is a major problem that limits overall efficiency and yield of crop. Priming of seed is an effective method for raising seed performance and improving tolerance of crops to abiotic stresses especially drought. The seeds of two local wheat cultivars (Kohistan-97 and Pasban-90) were soaked in distilled water or sodium selenate solutions of 25, 50, 75, and 100 μM for 1/2 or 1 h at 25 °C and later re-dried to their original moisture levels before sowing. One-hour priming significantly increased root length stress tolerance index, dry matter stress tolerance index, and total biomass of seedlings; however, no significant effect of changing duration of Se seed priming was observed on plant height stress tolerance index and shoot/root ratio. Among cultivars, Kohistan-97 was found to be more responsive to Se seed treatment as 1 h priming at 100 μM significantly increased its total biomass by 43 % as compared to control treatment. Although biomass of seedlings was not affected with Se seed priming under normal conditions, but it increased significantly with increase in rates of Se under drought stress conditions. One-hour priming at 75 μM increased the total sugar content and total free amino acids in both wheat cultivars. A more significant decrease in soluble proteins of seedlings was observed by 1 h priming than 1/2 h priming under drought stress conditions.
Sigma-Aldrich
Sodium selenate, ≥95% (elemental analysis)
Sigma-Aldrich
Sodium selenate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium selenate, BioXtra