Merck
  • Neuroprotective effects of oxymatrine against excitotoxicity partially through down-regulation of NR2B-containing NMDA receptors.
Neuroprotective effects of oxymatrine against excitotoxicity partially through down-regulation of NR2B-containing NMDA receptors.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2012-12-12)
Kun Zhang, Yu-jiao Li, Qi Yang, Oudeng Gerile, Le Yang, Xu-bo Li, Yan-yan Guo, Nan Zhang, Bin Feng, Shui-bing Liu, Ming-gao Zhao
ABSTRACT
Oxymatrine (OMT) is a major bioactive component derived from Sophora flavescens Ait (kushen), which is widely used in Chinese medicine. Recent studies have shown that it has neuroprotective effects; however, its underlying mechanisms remain unclear. We focus on the mechanisms of pharmacologic action in OMT by detecting its pharmacological properties against focal cerebral ischemia in vivo and NMDA-induced neurotoxicity in vitro. OMT prevented cerebral ischemic injury in mice induced via a 2 h middle cerebral artery occlusion and a 24 h reperfusion, in vivo. In vitro cultured neurons challenged with N-methyl-D-aspartate (NMDA, 200 μM) for 30 min showed significant decrease in the viability of neurons; however, OMT was able to protect neurons against induced neurotoxicity via NMDA exposure. Western blot analysis revealed that OMT decreased the expression of Bax and repaired the balance of pro- and anti-apoptotic proteins. Furthermore, OMT significantly reversed the up-regulation of NR2B and inhibited the calcium overload in the cultured neurons after challenging the NMDA. OMT showed partial protection in the cortical neurons via down-regulation of NR2B containing NMDA receptors and up-regulation of Bcl-2 family. Our results provide new insights into the development of natural therapeutic anti-oxidants against ischemia.