Merck
  • In chemico evaluation of skin metabolism: Investigation of eugenol and isoeugenol by electrochemistry coupled to liquid chromatography and mass spectrometry.
In chemico evaluation of skin metabolism: Investigation of eugenol and isoeugenol by electrochemistry coupled to liquid chromatography and mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-01-05)
Daniel Melles, Torsten Vielhaber, Anne Baumann, Raniero Zazzeroni, Uwe Karst
ABSTRACT
Skin sensitization is initiated by the modification of proteins located in the skin. After oxidative activation, eugenol and isoeugenol have the potential to modify skin proteins and therefore cause sensitization processes. Despite their known skin sensitizing properties, they are of common use in cosmetic products. According to the European Commission regulation No. 1223/2009, animal tests have to be banned for substances intended for cosmetic use. Therefore, alternative methods of investigation need to be developed for the approval of future substances. For this reason, eugenol and isoeugenol were selected as model substances to be investigated in a purely instrumental approach comprising electrochemistry, liquid chromatography and mass spectrometry. In the present work, reactive oxidation products of eugenol and isoeugenol were electrochemically generated. Reactive quinones and quinone methides were formed. Surprisingly, eugenol and isoeugenol differ significantly in their oxidation behaviour. Isoeugenol exhibits the formation of quinones and quinone methides of an alkylated and dealkylated species, respectively, whereas eugenol shows the formation of quinoid species only after dealkylation. Reactive quinoid species could be trapped with glutathione and the protein β-lactoglobulin A. The results are comparable to the ones with conventional animal studies in literature, which attribute the adverse effects of eugenol and isoeugenol to the formation of reactive quinones or quinone methides, which are reactive intermediates, able to react with proteins. Such species were successfully generated and investigated by the use of electrochemistry coupled to mass spectrometry. Above all, the investigation of adduct formation by the additional use of liquid chromatography allowed the assessment of the mechanism of oxidation, as it might happen in the skin. Both substances were proven to be trapped by the protein β-lactoglobulin A after electrochemical oxidation. However, isoeugenol formed the larger variety of adducts compared to eugenol.
Product Description
Supelco
Isoeugenol, analytical standard
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material