Merck
  • Purification and characterization of ovotransferrin from Crocodylus siamensis.
Purification and characterization of ovotransferrin from Crocodylus siamensis.
The protein journal (2013-01-19)
Sukanya Chaipayang, Napus Heamatorn, Likhid Keha, Sakda Daduang, Chomphunuch Songsiriritthigul, Prasan Swatsitang, Apisak Dhiravisit, Sompong Thammasirirak
ABSTRACT
Ovotransferrin (OTf) is the major glycoprotein in reptile egg whites. However, knowledge concerning its functional and biological properties remains limited. In this study, OTf from Crocodylus siamensis was purified and characterized. The proteins were precipitated with 80 % ammonium sulfate and then purified by anion exchange chromatography followed by hydrophobic interaction chromatography. The purified crocodile ovotransferrin (cOTf) had a molecular weight of 79 kDa. Analysis by two-dimensional polyacrylamide gel electrophoresis (2D-PAGE) indicated multiple isoforms of cOTf, which had isoelectric points ranging from 6.0 to 6.8. cOTf was N-linked glycosylated protein identified by using PNGase F deglycosylation technique. Optimal autoproteolysis of cOTf occurred under acidic conditions and pH values more than 5, which differs from that of OTf.
Sigma-Aldrich
Conalbumin from chicken egg white, suitable for cell culture, BioReagent
Sigma-Aldrich
Conalbumin from chicken egg white, Substantially iron-free
Sigma-Aldrich
Conalbumin from chicken egg white, Iron complex