Merck
  • Simultaneous determination of two-component isotherm parameters and lumped mass transfer coefficients in RPLC with the 0-1 model-inverse method.
Simultaneous determination of two-component isotherm parameters and lumped mass transfer coefficients in RPLC with the 0-1 model-inverse method.
Journal of separation science (2013-01-24)
Liang-Fei OuYang, Qian Liu, Heng Liang, Yuhua Shi, Junwei Shi, Xiaoguang Yin
ABSTRACT
The 0-1 model-inverse method of nonequilibrium nonlinear chromatography was developed to simultaneously determine the isotherm parameters and the lumped mass transfer coefficients of the two-component systems in RPLC. By comparing the simulated elution curves with experimental curves with regard to profiles and areas, the suitable isotherm parameters and the lumped mass transfer coefficients were obtained with the 0-1 model-inverse method. With a solute cell unit width of 1×10(-3) cm, the average errors of the peak areas were 0.178% for one component and -0.40% for two components, and the numerical diffusions of the 0-1 model for the contribution to band broadening may be negligible. In addition, the results showed that the lumped mass transfer coefficients decrease as the solute concentration increases. The 0-1 model-inverse method has not only the advantages of high calculation speed (less than 10 min for one-component systems or approximately 3 h for two-component systems using an ordinary computer) and high accuracy in simultaneously obtaining thermodynamic parameters and kinetic parameters of two-component systems, but this method also possesses the potential to optimally design and control the time-variant preparative chromatographic system due to the thermodynamic state recursion and the Lagrangian-Eulerian presentation of the 0-1 model.
Product Description
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material