Merck
  • A platform pathway for production of 3-hydroxyacids provides a biosynthetic route to 3-hydroxy-γ-butyrolactone.
A platform pathway for production of 3-hydroxyacids provides a biosynthetic route to 3-hydroxy-γ-butyrolactone.
Nature communications (2013-01-31)
Collin H Martin, Himanshu Dhamankar, Hsien-Chung Tseng, Micah J Sheppard, Christopher R Reisch, Kristala L J Prather
ABSTRACT
The replacement of petroleum feedstocks with biomass to produce platform chemicals requires the development of appropriate conversion technologies. 3-Hydroxy-γ-butyrolactone has been identified as one such chemical; however, there are no naturally occurring biosynthetic pathways for this molecule or its hydrolyzed form, 3,4-dihydroxybutyric acid. Here we design a novel pathway to produce various chiral 3-hydroxyacids, including 3,4-dihydroxybutyric acid, consisting of enzymes that condense two acyl-CoAs, stereospecifically reduce the resulting β-ketone and hydrolyze the CoA thioester to release the free acid. Acetyl-CoA serves as one substrate for the condensation reaction, whereas the second is produced intracellularly by a pathway enzyme that converts exogenously supplied organic acids. Feeding of butyrate, isobutyrate and glycolate results in the production of 3-hydroxyhexanoate, 3-hydroxy-4-methylvalerate and 3,4-dihydroxybutyric acid+3-hydroxy-γ-butyrolactone, respectively, molecules with potential uses in applications from materials to medicines. We also unexpectedly observe the condensation reaction resulting in the production of the 2,3-dihydroxybutyric acid isomer, a potential value-added monomer.
Product Description
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether