Merck
  • Management of atmospheric pollutants from waste incineration processes: the case of Bozen.
Management of atmospheric pollutants from waste incineration processes: the case of Bozen.
Waste management & research : the journal of the International Solid Wastes and Public Cleansing Association, ISWA (2013-02-01)
Marco Ragazzi, Werner Tirler, Giulio Angelucci, Dino Zardi, Elena Cristina Rada
ABSTRACT
This article presents the case study of a waste incinerator located in a region rich in natural and environmental resources, and close to the city of Bozen, where there are about 100,000 inhabitants. Local authorities paid special attention to the effect of the plant on human health and the surrounding environment. Indeed, among the measures adopted to control the emissions, in 2003 an automatic sampling system was installed specifically to monitor polychlorinated dibenzo-p-dioxin and polychlorinated dibenzofuran (PCDD/F) emissions during the complete operation time of the plant. The continuous sampling system was coupled directly to aerosol spectrometers for the determination of fine and ultra-fine particles in the emissions of the plant. The measurement results suggest that the waste incineration plant of Bozen is not a significant source of PCDD/F, or fine and ultra-fine particles. Immission measurements from other monitoring systems confirmed these results.
Product Description
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)