Merck
Helical deformation of the filament of a scroll wave.
Physical review. E, Statistical, nonlinear, and soft matter physics (2013-02-02)
Dennis Kupitz, Marcus J B Hauser
ABSTRACT
In three-dimensional excitable systems scroll waves may lose their originally straight shape through different instabilities. In experiments, the formation of zigzag-shaped or helical filaments is often observed. Such a deformation may be due to either a twist-induced instability or a 3D meandering instability. We performed a systematic study using a Belousov-Zhabotinsky (BZ) system with a vertical gradient of excitability and determined the necessary twist for the onset of undulation in the filament. Thus, we demonstrate that in the case of a BZ system with a gradient parallel to the filament, the deformation of the filament is induced by a twist-induced instability.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%