Merck
  • Induction of sister chromatid exchange by acrylamide and glycidamide in human lymphocytes: role of polymorphisms in detoxification and DNA-repair genes in the genotoxicity of glycidamide.
Induction of sister chromatid exchange by acrylamide and glycidamide in human lymphocytes: role of polymorphisms in detoxification and DNA-repair genes in the genotoxicity of glycidamide.
Mutation research (2013-02-05)
Marta Pingarilho, Nuno G Oliveira, Célia Martins, Bruno Costa Gomes, Ana Sofia Fernandes, Vanda Martins, Anatália Labilloy, João Pereira de Lima, José Rueff, Jorge Francisco Gaspar
ABSTRACT
Acrylamide (AA) is a probable human carcinogen generated in carbohydrate-rich foodstuffs upon heating. Glycidamide (GA), formed via epoxidation, presumably mediated by cytochrome P450 2E1, is considered to be the active metabolite that plays a central role in the genotoxicity of AA. The aim of this work was to evaluate the cytogenetic damage induced by AA and GA in cultured human lymphocytes by use of the sister chromatid exchange (SCE) assay. Furthermore, this report addresses the role of individual genetic polymorphisms in key genes involved in detoxification and DNA-repair pathways (BER, NER, HRR and NHEJ) on the induction of SCE by GA. While AA induced the number of SCE/metaphase only slightly, especially for the highest concentration tested (2000μM), GA markedly induced SCEs in a concentration-dependent manner up to concentrations of 750μM, leading to an increase in SCEs of up to about 10-fold compared with controls. By combining DNA damage in GA-treated lymphocytes and data on polymorphisms, associations between the induction of SCEs with GSTP1 (Ile105Val) and GSTA2 (Glu210Ala) genotypes are suggested.
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Supelco
Glycidamide, analytical standard
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered