Merck
  • Characterization of an isoeugenol monooxygenase (Iem) from Pseudomonas nitroreducens Jin1 that transforms isoeugenol to vanillin.
Characterization of an isoeugenol monooxygenase (Iem) from Pseudomonas nitroreducens Jin1 that transforms isoeugenol to vanillin.
Bioscience, biotechnology, and biochemistry (2013-02-09)
Ji-Young Ryu, Jiyoung Seo, Sunhwa Park, Joong-Hoon Ahn, Youhoon Chong, Michael J Sadowsky, Hor-Gil Hur
ABSTRACT
The isoeugenol monooxygenase (iem) gene from Pseudomonas nitroreducens Jin1, responsible for the conversion of isoeugenol to vanillin, was cloned and overexpressed in Escherichia coli. The purified Iem had a predicted molecular mass of 54 kDa. The V(max), K(M), and k(cat) values for it, using isoeugenol as substrate, were 4.2 µmol vanillin min(-1) mg(-1) of protein, 120 µM, and 3.8 s(-1), respectively. Maximum substrate turnover for Iem occurred at 30 °C and pH 9.0. An (18)Oxygen-labeling experiment revealed that oxidative cleavage of isoeugenol by Iem was catalyzed via a monooxygenation reaction, and that incorporation of the oxygen atom from O(2) into vanillin was preferable to incorporation from water. While the catalytic activity of Iem, as prepared, did not require the addition of any organic or metal cofactor, ICP-MS analysis showed 0.7 mol of iron per mol of Iem. Moreover site-directed mutagenesis of Iem of four conserved histidine residues individually, His(167), His(218), His(282), and His(471), which appear to be involved in ligand bonding with Fe(2+), resulted in a loss of activity. Enzyme activity was not appreciably influenced by preincubation of Iem with high concentrations of chelators, suggesting that iron is tightly bound. Iem has an iron-mediated mechanism that is widely spread among the three domains of life.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isoeugenol, mixture of cis and trans, 99%, FG
Supelco
Isoeugenol, analytical standard
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG