Merck
  • Thyroglobulin in washout fluid from lymph node fine-needle aspiration biopsy in papillary thyroid cancer: large-scale validation of the cutoff value to determine malignancy and evaluation of discrepant results.
Thyroglobulin in washout fluid from lymph node fine-needle aspiration biopsy in papillary thyroid cancer: large-scale validation of the cutoff value to determine malignancy and evaluation of discrepant results.
The Journal of clinical endocrinology and metabolism (2013-02-09)
Jae Hoon Moon, Yong Il Kim, Jung Ah Lim, Hoon Sung Choi, Sun Wook Cho, Kyung Won Kim, Hyo Jin Park, Jin Chul Paeng, Young Joo Park, Ka Hee Yi, Do Joon Park, Sang Eun Kim, June-Key Chung
ABSTRACT
There are still some controversies regarding the cutoff value and the influential factors of thyroglobulin (Tg) concentration in washout fluid from fine-needle aspiration (FNA) biopsy (FNA-Tg) on cervical lymph nodes (LNs) in patients with papillary thyroid cancer (PTC). Our aims were to validate the cutoff value of FNA-Tg in diagnosing malignant LNs on a large scale and to investigate the influential factors that could result in the discrepancy between the final diagnosis and FNA-Tg. We conducted a retrospective cohort study based on hospital records with 528 cases of FNA-Tg measurement from 419 PTC patients. The cutoff value of FNA-Tg was obtained from receiver operating characteristic analysis with final diagnosis. Binary logistic regression analysis was performed to investigate the influential factors. In the final diagnosis, 190 LNs were malignant, and 338 LNs were benign. The median FNA-Tg was 521.2 (3676.8) ng/mL in malignant LNs, and 0.1 (0.2) ng/mL in benign LNs. The optimal cutoff value of FNA-Tg in distinguishing malignant LNs from benign LNs was 1.0 ng/mL (sensitivity, 93.2%; specificity, 95.9%) in all cases. Combining FNA-Tg and FNA cytology showed superior diagnostic power (sensitivity, 98.4%; specificity, 94.4%) when compared with diagnostic strategy using either FNA cytology or FNA-Tg alone. FNA-Tg, serum TSH, and serum Tg were higher in nonthyroidectomized patients than in thyroidectomized patients (P < .001, respectively). FNA-Tg was correlated with serum TSH and Tg levels (P < .001, respectively), and binary logistic regression analysis showed that serum TSH suppression and serum Tg presence independently affected the diagnosis made by FNA-Tg. Our results validated 1.0 ng/mL of FNA-Tg as a cutoff value for diagnosing LN metastasis of PTC and suggested that serum TSH suppression and serum Tg presence should be considered in diagnosing LN malignancy with FNA-Tg in PTC patients.
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder
Sigma-Aldrich
Thyroglobulin human