Merck
  • Neighboring effect in fragmentation pathways of cage guanylhydrazones in the gas phase.
Neighboring effect in fragmentation pathways of cage guanylhydrazones in the gas phase.
The journal of physical chemistry. A (2013-02-19)
Marina Šekutor, Zoran Glasovac, Kata Mlinarić-Majerski
ABSTRACT
ESI-MS/MS investigation of the mono- and bis(guanylhydrazone) derivatives 1-5 based on adamantane and pentacycloundecane (PCU) skeleton was described. Elimination of neutral guanidine is the most abundant reaction channel in the case of 2,4-adamantyl and PCU derivatives 4 and 5, while the elimination of CH2N2 fragment is preferred for other compounds. This was attributed to the cage opening of adamantane or PCU skeletons in the former case leading to the formation of the cyclohexyl- or cyclopropylcarbinyl carbocation stabilized by the conjugation with the guanylhydrazone subunit. The main fragmentation pathways observed experimentally were analyzed by using DFT calculations. All investigated bis(guanylhydrazone)s formed dications and their abundances were found to be proportional to the interguanidine distance in the considered ions. Calculation of the first and the second proton affinities supported qualitative interpretation of the dication abundance. Close contact of two guanidine subunits is thus confirmed to be crucial in determining preferential fragmentation pathway and to suppress formation of the dication.
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Adamantane, ≥99%
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)