Merck
  • Molecular structural, non-linear optical, second order perturbation and Fukui studies of Indole-3-Aldehyde using density functional calculations.
Molecular structural, non-linear optical, second order perturbation and Fukui studies of Indole-3-Aldehyde using density functional calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-19)
S Muthu, J Uma Maheswari, Tom Sundius
ABSTRACT
Indole-3-Aldehyde is a new organic non-linear material having good second harmonic generation. The optimized molecular geometry, harmonic vibrational frequencies, infrared intensities of Indole-3-Aldehyde (I3A, C9H7NO) in the ground state were carried out by using density functional theory (B3LYP) method with 6-31G(d,p) basis set. A detailed interpretation of the infrared spectrum of Indole-3-Aldehyde is reported. The vibrational frequencies are calculated and compared with experimental FT-IR spectra. The theoretical spectrograms of FT-IR of the title compound have been constructed in addition, theoretical information like ONIOM, potential energy surface, NBO, and Fukui function are also calculated. Unambiguous vibrational assignment of all the fundamentals was made using the potential energy distribution.
Sigma-Aldrich
Indole-3-carboxaldehyde, 97%