Merck
  • Acupuncture in patients with seasonal allergic rhinitis: a randomized trial.
Acupuncture in patients with seasonal allergic rhinitis: a randomized trial.
Annals of internal medicine (2013-02-20)
Benno Brinkhaus, Miriam Ortiz, Claudia M Witt, Stephanie Roll, Klaus Linde, Florian Pfab, Bodo Niggemann, Josef Hummelsberger, András Treszl, Johannes Ring, Torsten Zuberbier, Karl Wegscheider, Stefan N Willich
ABSTRACT
Chinese translation Acupuncture is frequently used to treat seasonal allergic rhinitis (SAR) despite limited scientific evidence. To evaluate the effects of acupuncture in patients with SAR. Randomized, controlled multicenter trial. (ClinicalTrials.gov: NCT00610584) 46 specialized physicians in 6 hospital clinics and 32 private outpatient clinics. 422 persons with SAR and IgE sensitization to birch and grass pollen. Acupuncture plus rescue medication (RM) (cetirizine) (n = 212), sham acupuncture plus RM (n = 102), or RM alone (n = 108). Twelve treatments were provided over 8 weeks in the first year. Changes in the Rhinitis Quality of Life Questionnaire (RQLQ) overall score and the RM score (RMS) from baseline to weeks 7 and 8 and week 16 in the first year and week 8 in the second year after randomization, with predefined noninferiority margins of -0.5 point (RQLQ) and -1.5 points (RMS). Compared with sham acupuncture and with RM, acupuncture was associated with improvement in RQLQ score (sham vs. acupuncture mean difference, 0.5 point [97.5% CI, 0.2 to 0.8 point; P < 0.001]; RM vs. acupuncture mean difference, 0.7 point [97.5% CI, 0.4 to 1.0 point; P < 0.001]) and RMS (sham vs. acupuncture mean difference, 1.1 points [97.5% CI, 0.4 to 1.9 points; P < 0.001]; RM vs. acupuncture mean difference, 1.5 points [97.5% CI, 0.8 to 2.2 points; P < 0.001]). There were no differences after 16 weeks in the first year. After the 8-week follow-up phase in the second year, small improvements favoring real acupuncture over the sham procedure were noted (RQLQ mean difference, 0.3 point [95% CI, 0.03 to 0.6 point; P = 0.032]; RMS mean difference, 1.0 point [95% CI, 0.2 to 1.9 points; P = 0.018]). The study was not powered to detect rare adverse events, and the RQLQ and RMS values were low at baseline. Acupuncture led to statistically significant improvements in disease-specific quality of life and antihistamine use measures after 8 weeks of treatment compared with sham acupuncture and with RM alone, but the improvements may not be clinically significant.
Sigma-Aldrich
Cetirizin dihydrochloride, ≥98.0% (HPLC)
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard