Merck
  • A derivative photoelectrochemical sensing platform for 4-nitrophenolate contained organophosphates pesticide based on carboxylated perylene sensitized nano-TiO2.
A derivative photoelectrochemical sensing platform for 4-nitrophenolate contained organophosphates pesticide based on carboxylated perylene sensitized nano-TiO2.
Analytica chimica acta (2013-02-23)
Hongbo Li, Jing Li, Qin Xu, Zhanjun Yang, Xiaoya Hu
ABSTRACT
A novel visible light sensitized photoelectrochemical sensing platform was constructed based on the perylene-3,4,9,10-tetracarboxylic acid/titanium dioxide (PTCA/TiO(2)) heterojunction as the photoelectric beacon. PTCA was synthesized via facile steps of hydrolysis and neutralization reaction, and then the PTCA/TiO(2) heterojunction was easily prepared by coating PTCA on nano-TiO(2) surface. The resulting photoelectric beacon was characterized by transmission electron microscope, scanning electron microscopy, X-ray diffractometry, FTIR spectroscopy, and ultraviolet and visible spectrophotometer. Using parathion-methyl as a model, after a simple hydrolyzation process, p-nitrophenol as the hydrolysate of parathion-methyl could be obtained, the fabricated derivative photoelectrochemical sensor showed good performances with a rapid response, instrument simple and portable, low detection limit (0.08 nmol L(-1)) at a signal-to-noise ratio of 3, and good selectivity against other pesticides and possible interferences. It had been successfully applied to the detection of parathion-methyl in green vegetables and the results agreed well with that by GC-MS. This strategy not only extends the application of PTCA, but also presents a simple, economic and novel methodology for photoelectrochemical sensing.
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
Parathion-methyl, PESTANAL®, analytical standard
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%