Merck
  • Akt kinase targets the association of CBP with histone H3 to regulate the acetylation of lysine K18.
Akt kinase targets the association of CBP with histone H3 to regulate the acetylation of lysine K18.
FEBS letters (2013-02-26)
Yan Liu, Zhao-bin Xing, Jing-hua Zhang, Yue Fang
ABSTRACT
CREB binding protein (CBP) is an acetyltransferase that plays an important role in many biological processes. Here, we show that Akt phosphorylates CBP at threonine 1871 and suppresses its acetyltransferase activity by impeding the binding of CBP to histone H3, which results in a decrease in lysine K18 acetylation and dysregulation of target genes. Our results demonstrate that Akt regulates acetyltransferase activity through CBP phosphorylation, which may contribute to tumorigenesis.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material