Merck
  • Associations of ambient hydrogen sulfide exposure with self-reported asthma and asthma symptoms.
Associations of ambient hydrogen sulfide exposure with self-reported asthma and asthma symptoms.
Environmental research (2013-03-05)
Michael N Bates, Nick Garrett, Julian Crane, John R Balmes
ABSTRACT
Whether long-term, low-level hydrogen sulfide (H2S) gas is a cause of health effects, including asthma, is uncertain. Rotorua city, New Zealand, has the largest population exposed, from geothermal sources, to relatively high ambient levels of H2S. In a cross-sectional study, the authors investigated associations with asthma in this population. A total of 1637 adults, aged 18-65 years, were enrolled during 2008-2010. Residences and workplaces were geocoded. H2S exposures at homes and workplaces were estimated using city-wide networks of passive H2S samplers and kriging to create exposure surfaces. Exposure metrics were based on (1) time-weighted exposures at home and work; and (2) the maximum exposure (home or work). Exposure estimates were entered as quartiles into regression models, with covariate data. Neither exposure metric showed evidence of increased asthma risk from H2S. However, some suggestion of exposure-related reduced risks for diagnosed asthma and asthma symptoms, particularly wheezing during the last 12 months, emerged. With the maximum exposure metric, the prevalence ratio for wheeze in the highest exposure quartile was 0.80 (0.65, 0.99) and, for current asthma treatment, 0.75 (0.52, 1.08). There was no evidence that this was caused by a "survivor effect". The study provided no evidence that asthma risk increases with H2S exposure. Suggestions of a reduced risk in the higher exposure areas are consistent with recent evidence that H2S has signaling functions in the body, including induction of smooth muscle relaxation and reduction of inflammation. Study limitations, including possible confounding, preclude definitive conclusions.
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%