Merck
Structures and reaction mechanisms of GTP cyclohydrolases.
IUBMB life (2013-03-05)
Tobias Gräwert, Markus Fischer, Adelbert Bacher
ABSTRACT
GTP cyclohydrolases generate the first committed intermediates for the biosynthesis of certain vitamins/cofactors (folic acid, riboflavin, deazaflavin, and tetrahydrobiopterin), deazapurine antibiotics, some t-RNA bases (queuosine, archaeosine), and the phytotoxin, toxoflavin. They depend on divalent cations for hydrolytic opening of the imidazole ring of the substrate, guanosine triphosphate (GTP). Surprisingly, the ring opening reaction is not the rate-limiting step for GTP cyclohydrolases I and II whose mechanism have been studied in some detail. GTP cyclohydrolase I, Ib, and II are potential targets for novel anti-infectives. Genetic factors modulating the activity of human GTP cyclohydrolase are highly pleiotropic, since the signal transponders whose biosyntheses require their participation (nitric oxide, catecholamines) impact a very wide range of physiological phenomena. Recent studies suggest that human GTP cyclohydrolase may become an oncology target.
Product Description
Supelco
Riboflavin (B2), analytical standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
6-Biopterin, ≥97%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%