Merck
  • Mast cell expression of the serotonin1A receptor in guinea pig and human intestine.
Mast cell expression of the serotonin1A receptor in guinea pig and human intestine.
American journal of physiology. Gastrointestinal and liver physiology (2013-03-23)
Guo-Du Wang, Xi-Yu Wang, Fei Zou, Meihua Qu, Sumei Liu, Guijun Fei, Yun Xia, Bradley J Needleman, Dean J Mikami, Jackie D Wood
ABSTRACT
Serotonin [5-hydroxytryptamine (5-HT)] is released from enterochromaffin cells in the mucosa of the small intestine. We tested a hypothesis that elevation of 5-HT in the environment of enteric mast cells might degranulate the mast cells and release mediators that become paracrine signals to the enteric nervous system, spinal afferents, and secretory glands. Western blotting, immunofluorescence, ELISA, and pharmacological analysis were used to study expression of 5-HT receptors by mast cells in the small intestine and action of 5-HT to degranulate the mast cells and release histamine in guinea pig small intestine and segments of human jejunum discarded during Roux-en-Y gastric bypass surgeries. Mast cells in human and guinea pig preparations expressed the 5-HT1A receptor. ELISA detected spontaneous release of histamine in guinea pig and human preparations. The selective 5-HT1A receptor agonist 8-hydroxy-PIPAT evoked release of histamine. A selective 5-HT1A receptor antagonist, WAY-100135, suppressed stimulation of histamine release by 5-HT or 8-hydroxy-PIPAT. Mast cell-stabilizing drugs, doxantrazole and cromolyn sodium, suppressed the release of histamine evoked by 5-HT or 8-hydroxy-PIPAT in guinea pig and human preparations. Our results support the hypothesis that serotonergic degranulation of enteric mast cells and release of preformed mediators, including histamine, are mediated by the 5-HT1A serotonergic receptor. Association of 5-HT with the pathophysiology of functional gastrointestinal disorders (e.g., irritable bowel syndrome) underlies a question of whether selective 5-HT1A receptor antagonists might have therapeutic application in disorders of this nature.
Sigma-Aldrich
Ketotifen fumarate salt
Sigma-Aldrich
Cromolyn sodium salt, ≥95%
Sigma-Aldrich
Donkey Anti-Guinea Pig IgG Antibody, FITC conjugate, Species Adsorbed, Chemicon®, from donkey