Merck
  • Polycyclic aromatic hydrocarbon removal from petroleum sludge cake using thermal treatment with additives.
Polycyclic aromatic hydrocarbon removal from petroleum sludge cake using thermal treatment with additives.
Environmental technology (2013-03-28)
Edward Nixon Pakpahan, Mohamed Hasnain Isa, Shamsul Rahman Mohamed Kutty, Somporn Chantara, Wan Wiriya
ABSTRACT
Petroleum sludge is a hazardous waste that contains various organic compounds including polycyclic aromatic hydrocarbons (PAHs) which have carcinogenic-mutagenic and toxic characteristics. This study focuses on the thermal treatment (indirect heating) of petroleum sludge cake for PAH degradation at 250, 450, and 650 degrees C using Ca(OH)2 + NaHCO3 as an additive. The treatment was conducted in a rotary drum electric heater. All experiments were carried out in triplicate. Concentrations of the 16 priority PAHs in gas (absorbed on Amberlite XAD-4 adsorbent), particulate (on quartz filter) and residue phases were determined using gas chromatography-mass spectrometry (GC-MS). The samples were extracted with acetonitrile by ultra-sonication prior to GC-MS analysis. The use of additive was beneficial and a temperature of 450 degrees C was suitable for PAH degradation. Low levels of PAH emissions, particularly carcinogenic PAH and toxic equivalent concentration (sigma TEC), were observed in gas, particulate and residue phases after treatment.
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium hydroxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium hydroxide, ACS reagent, ≥95.0%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C