Merck
  • Behavioral effects of cocaine mediated by nitric oxide-GAPDH transcriptional signaling.
Behavioral effects of cocaine mediated by nitric oxide-GAPDH transcriptional signaling.
Neuron (2013-05-31)
Risheng Xu, Anthony V Serritella, Tanusree Sen, Justin M Farook, Thomas W Sedlak, Jay Baraban, Solomon H Snyder, Nilkantha Sen
ABSTRACT
Cocaine's behavioral-stimulant effects derive from potentiation of synaptic signaling by dopamine and serotonin leading to transcriptional alterations in postsynaptic cells. We report that a signaling cascade involving nitric oxide (NO) and glyceraldehyde-3-phosphate dehydrogenase (GAPDH) mediates cocaine's transcriptional and behavioral actions. Lower, behavioral-stimulant doses enhance the cAMP response element-binding (CREB) signaling system, while higher, neurotoxic doses stimulate the p53 cytotoxic system. The drug CGP3466B, which potently and selectively blocks GAPDH nitrosylation and GAPDH-Siah binding, prevents these actions as well as behavioral effects of cocaine providing a strategy for anticocaine therapy.
Sigma-Aldrich
Cocaine free base
Supelco
Cocaine hydrochloride solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Supelco
Cocaine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cocaine hydrochloride