Merck
  • Thiol-based antioxidant supplementation alters human skeletal muscle signaling and attenuates its inflammatory response and recovery after intense eccentric exercise.
Thiol-based antioxidant supplementation alters human skeletal muscle signaling and attenuates its inflammatory response and recovery after intense eccentric exercise.
The American journal of clinical nutrition (2013-05-31)
Yannis Michailidis, Leonidas G Karagounis, Gerasimos Terzis, Athanasios Z Jamurtas, Kontantinos Spengos, Dimitrios Tsoukas, Athanasios Chatzinikolaou, Dimitrios Mandalidis, Renae J Stefanetti, Ioannis Papassotiriou, Spyros Athanasopoulos, John A Hawley, Aaron P Russell, Ioannis G Fatouros
ABSTRACT
The major thiol-disulfide couple of reduced glutathione (GSH) and oxidized glutathione is a key regulator of major transcriptional pathways regulating aseptic inflammation and recovery of skeletal muscle after aseptic injury. Antioxidant supplementation may hamper exercise-induced cellular adaptations. The objective was to examine how thiol-based antioxidant supplementation affects skeletal muscle's performance and redox-sensitive signaling during the inflammatory and repair phases associated with exercise-induced microtrauma. In a double-blind, crossover design, 10 men received placebo or N-acetylcysteine (NAC; 20 mg · kg(-1) · d(-1)) after muscle-damaging exercise (300 eccentric contractions). In each trial, muscle performance was measured at baseline, after exercise, 2 h after exercise, and daily for 8 consecutive days. Muscle biopsy samples from vastus lateralis and blood samples were collected before exercise and 2 h, 2 d, and 8 d after exercise. NAC attenuated the elevation of inflammatory markers of muscle damage (creatine kinase activity, C-reactive protein, proinflammatory cytokines), nuclear factor κB phosphorylation, and the decrease in strength during the first 2 d of recovery. NAC also blunted the increase in phosphorylation of protein kinase B, mammalian target of rapamycin, p70 ribosomal S6 kinase, ribosomal protein S6, and mitogen activated protein kinase p38 at 2 and 8 d after exercise. NAC also abolished the increase in myogenic determination factor and reduced tumor necrosis factor-α 8 d after exercise. Performance was completely recovered only in the placebo group. Although thiol-based antioxidant supplementation enhances GSH availability in skeletal muscle, it disrupts the skeletal muscle inflammatory response and repair capability, potentially because of a blunted activation of redox-sensitive signaling pathways. This trial was registered at clinicaltrials.gov as NCT01778309.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein