Merck
  • [Homocysteine levels and platelet aggregation in patients with cerebral circulatory disorders].
[Homocysteine levels and platelet aggregation in patients with cerebral circulatory disorders].
Terapevticheskii arkhiv (2013-05-31)
Z Sabaliauskiene, P Gribauskas, V Gaigalaite, J Ptasekas, V Ojeraitene, D Kalibatene
ABSTRACT
To reveal platelet functional changes manifesting as abnormal rate of platelet aggregation in patients with acute cerebral circulatory disorders (ischemic stroke, transient ischemic attack), to evaluate the effect of homocysteine (Hc) on platelet aggregation, depending on the degree of cerebral blood flow disturbances, patients' gender and age, and to estimate the diagnostic value of their associations. 50 patients aged 33 to 98 years (mean age, 63.7 +/- 2.1 years; 20 men and 30 women) with acute cerebral circulatory disorders (18 with transient ischemic attack and 32 with ischemic stroke) were examined. The diagnosis was verified by the results of computed tomography and other clinical examinations. Adenosine diphosphate-, epinephrine-, and collagen-induced platelet aggregation was assessed in platelet-rich plasma; serum Hc concentrations were also studied. Comparison of platelet aggregation and Hc concentration revealed a statistically significant correlation between platelet aggregation with collagen and elevated Hc levels (r = 0.376; p < 0.01). There was no statistically significant correlation with other inducers. Blood Hc concentrations increased with advancing age (r = 0.357; p = 0.015). No statistically significant correlation was found between age and platelet aggregation (p > 0.05). The levels of Hc were statistically significantly higher as cerebral circulatory disorders progressed. There was a statistically significantly relationship between collagen-induced platelet aggregation and Hc concentration. The Hc levels were statistically significantly related to the severity of cerebral circulatory disorders and age.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder