Merck
  • The treatment of Wilson's disease, a rare genetic disorder of copper metabolism.
The treatment of Wilson's disease, a rare genetic disorder of copper metabolism.
Science progress (2013-06-07)
Rupert Purchase
ABSTRACT
Wilson's disease is a rare autosomal recessive disease characterised by the deposition of copper in the brain, liver; cornea, and other organs. The overload of copper inevitably leads to progressive liver and neurological dysfunction. Copper overload in patients with Wilson's disease is caused by impairment to the biliary route for excretion of dietary copper A combination of neurological, psychiatric and hepatic symptoms can make the diagnosis of Wilson's disease challenging. Most symptoms appear in the second and third decades of life. The disease affects between one in 30,000 and one in 100,000 individuals, and is fatal if left untreated. Five drugs are currently available to treat Wilson's disease: British Anti-Lewisite; D-penicillamine; trientine; zinc sulfate or acetate; and ammonium tetrathiomolybdate. Each drug can reduce copper levels and/or transform copper into a metabolically inert and unavailable form in the patient. The discovery and introduction of these five drugs owes more to the inspiration of a few dedicated physicians and agricultural scientists than to the resources of the pharmaceutical industry.
Sigma-Aldrich
D-Penicillamine, 98-101%
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Triethylenetetramine, ≥97.0% (T)
Sigma-Aldrich
Triethylenetetramine, technical grade, 60%
Sigma-Aldrich
Zinc sulfate solution, BioUltra, Molecular Biology, 2.0 M in H2O
Sigma-Aldrich
Zinc sulfate solution, 0.3 N
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis