Merck
  • Revascularization/Regeneration performed in immature molars: case reports.
Revascularization/Regeneration performed in immature molars: case reports.
The Journal of clinical pediatric dentistry (2013-07-17)
I S Sönmez, A Akbay Oba, M Erkmen Almaz
ABSTRACT
These 3 case reports the outcome of revascularization treatment in necrotic immature molars. During treatment, a tri antibiotic mix was used to disinfect the pulp for 2 weeks. Then a blood clot was created in the canal, over which mineral trioxide aggregate was placed. After 24 months, the immature molars showed continuation of root development. The patients were asymptomatic, no sinus tracts were evident and apical periodontitis was resolved Results from these cases show that revascularization/regeneration using 3Mix-MP method could be effective for managing immature permanent molar teeth with pulpal necrosis.
Product Description
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, ≤50 nm particle size
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Supelco
Zinc oxide, analytical standard
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Minocycline hydrochloride, powder
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)