Merck
  • Building nanoSPR biosensor systems based on gold magnetic composite nanoparticles.
Building nanoSPR biosensor systems based on gold magnetic composite nanoparticles.
Journal of nanoscience and nanotechnology (2013-07-26)
Dong Yang, Jianzhong Ma, Mingli Peng, Qinlu Zhang, Yanling Luo, Wenli Hui, Tianbo Jin, Yali Cui
ABSTRACT
Composite Fe3O4/Au nanoparticles are attracting considerable interest in developing visual and specific detection of biomolecular due to their unique physical and chemical properties. Here, two localize surface plasmon resonance (LSPR) probes based on Fe3O4/Au nanoparticles (nanoSPR biosensors) were fabricated by exploring 11-mercaptoundecanoic acid (MUA) and poly acrylic acid (PAA) as surface modification agents and subsequently conjugating rabbit IgG with the modified particles' surface. Comparative investigations showed the differences between MUA-particles and PAA-particles, as well as sensitivity of the two as-prepared nanoSPR biosensors when used in target goat anti-rabbit IgG colorimetric detection. Particles coated with PAA were in a better dispersion and showed an ionic independent stability, indicating PAA-particles have a potential application in colorimetric detection. In contrast, the MUA-particle probes revealed a higher sensitivity in SPR detection (50 nmol/L), and further kinetic studies showed the reactions between probes and target followed the second order and the reaction rate of MUA-probes was twice the rate of PAA-probes at the same temperature and condition. Such proof-of concept works reported here demonstrated that the protocol to build nanoSPR biosensors was favored in developing molecular probes, and the novel composite nanoparticles might serve as ideal probes for sensitive, selective and real time detection.
Sigma-Aldrich
Poly(acrylic acid)
Supelco
Poly(acrylic acid sodium salt), analytical standard, suitable for gel permeation chromatography (GPC), 1,250
Supelco
Poly(acrylic acid sodium salt), analytical standard, suitable for gel permeation chromatography (GPC), 16,000
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass cover slip, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å