Merck
[Cocaine addiction].
Revue medicale de Liege (2013-07-31)
W Pitchot, G Scantamburlo, E Pinto, L Karila
ABSTRACT
Cocaine is the second most commonly used illicit drug after cannabis in the general population. Cocaine is a powerful stimulating agent of the central nervous system and a highly addictogenic drug. Somatic and psychiatric consequences of cocaine addiction are major and clinically relevant. The increasing consumption of cocaine and the importance of its consequences justify an update of our knowledge about cocaine addiction.
Product Description
Supelco
Cocaine hydrochloride solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Sigma-Aldrich
Cocaine hydrochloride
Supelco
Cocaine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cocaine free base