Merck
  • Sustained carbon uptake and storage following moderate disturbance in a Great Lakes forest.
Sustained carbon uptake and storage following moderate disturbance in a Great Lakes forest.
Ecological applications : a publication of the Ecological Society of America (2013-08-24)
Christopher M Gough, Brady S Hardiman, Lucas E Nave, Gil Bohrer, Kyle D Maurer, Christoph S Vogel, Knute J Nadelhoffer, Peter S Curtis
ABSTRACT
Carbon (C) uptake rates in many forests are sustained, or decline only briefly, following disturbances that partially defoliate the canopy. The mechanisms supporting such functional resistance to moderate forest disturbance are largely unknown. We used a large-scale experiment, in which > 6700 Populus (aspen) and Betula (birch) trees were stem-girdled within a 39-ha area, to identify mechanisms sustaining C uptake through partial canopy defoliation. The Forest Accelerated Succession Experiment in northern Michigan, USA, employs a suite of C-cycling measurements within paired treatment and control meteorological flux tower footprints. We found that enhancement of canopy light-use efficiency and maintenance of light absorption maintained net ecosystem production (NEP) and aboveground wood net primary production (NPP) when leaf-area index (LAI) of the treatment forest temporarily declined by nearly half its maximum value. In the year following peak defoliation, redistribution of nitrogen (N) in the treatment forest from senescent early successional aspen and birch to non-girdled later successional species facilitated the recovery of total LAI to pre-disturbance levels. Sustained canopy physiological competency following disturbance coincided with a downward shift in maximum canopy height, indicating that compensatory photosynthetic C uptake by undisturbed, later successional subdominant and subcanopy vegetation supported C-uptake resistance to disturbance. These findings have implications for ecosystem management and modeling, demonstrating that forests may tolerate considerable leaf-area losses without diminishing rates of C uptake. We conclude that the resistance of C uptake to moderate disturbance depends not only on replacement of lost leaf area, but also on rapid compensatory photosynthetic C uptake during defoliation by emerging later successional species.
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Supelco
Activated charcoal, puriss. p.a., powder
Supelco
Activated charcoal, powder
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular