Merck
  • Rat selection test with respect to laxative activity induced by barbaloin.
Rat selection test with respect to laxative activity induced by barbaloin.
Biological & pharmaceutical bulletin (1993-10-01)
Y Ishii, H Tanizawa, Y Takino
ABSTRACT
As individual differences in sensitivity in the laxative activity of barbaloin were observed in rats, a selection test for barbaloin (31.1 mg/kg, p.o.) was carried out between 1989 and 1991. The annual percentages of rats exhibiting a reaction positive to barbaloin were almost constant (about 57%). We propose that rats positive to barbaloin should be selected for studies on the laxative effect of barbaloin.
Sigma-Aldrich
Aloin, from Aloe barbadensis Miller leaves, ≥97%
Sigma-Aldrich
Aloin, from Curacao aloe, ~50%