Merck
  • Reversible addition-fragmentation chain transfer polymerization of N-isopropylacrylamide: a comparison between a conventional and a fast initiator.
Reversible addition-fragmentation chain transfer polymerization of N-isopropylacrylamide: a comparison between a conventional and a fast initiator.
The journal of physical chemistry. B (2007-09-07)
Hassen Bouchékif, Ravin Narain
ABSTRACT
The reversible addition-fragmentation chain transfer (RAFT) polymerization of N-isopropylacrylamide (NIPAM) was studied to determine the reasons for deviation of experimental molecular weights to lower molecular weight at high monomer conversion when S-1-dodecyl-S-(alpha,alpha'-dimethyl-alpha' '-acetic acid)trithiocarbonate (CTAm) and S,S-bis(alpha,alpha'-dimethyl-alpha' '-acetic acid)trithiocarbonate (CTAd) were used as RAFT agents at 65 degrees C. For this purpose, experiments were performed in N,N'-dimethylformamide (DMF) at the NIPAM/CTA ratio of 200 with initiators capable of yielding fast and slow initiation, respectively by photochemical and thermal process, either at ambient temperature or at 65 degrees C. When the polymerization of NIPAM was conducted under these conditions with Irgacure-2959 (IRGC) as photoinitiator, a continuing supply of primary radicals by incremental initiator addition was required to achieve reasonably high conversion. This effect was also apparent by the loss of linearity of the first-order kinetic plot with a conventional initiator (4,4-azobis(4-cyanovaleric acid) (ACVA) as azo-initiator, 10h (t1/2) decomposition at 65 degrees C) indicating that steady-state concentration of the macroradical decreases significantly with the initiator consumption. Nevertheless, polymers with predictable number-average molecular weight Mn (i.e., based on [monomer]/([CTA] + [initiator]) ratio) and narrow polydispersities were obtained (PDIs < 1.2) with CTAm indicating that the process of chain growth was controlled. When CTAd was used, instead of CTAm, the polymers obtained were characterized by a larger polydispersity (1.2 < PDIs < 1.3). The so-called "living steady-state concentration" in chain equilibration together with the linear dependence of Mn vs conversion was observed only when the 200/1 NIPAM/CTA mixture in DMF was subjected to a permanent photoirradiation at 65 degrees C. With ACVA, the deviation of the experimentally measured molecular weights at high conversion was accounted for by the simultaneous self-initiated polymerization of NIPAM with the controlled process in the presence of CTA at 65 degrees C. Similar drift from the linear dependence Mn vs conversion was also observed at 65 degrees C when a significant number of low molecular weight polymer chains were generated intentionally by photodecomposition of IRGC.
Sigma-Aldrich
4,4′-Azobis(4-cyanovaleric acid), ≥98.0% (T)