Merck
  • Evaluation of cytotoxicity of 3-mercaptopropionic acid-modified quantum dots on Medicago sativa cells and tissues.
Evaluation of cytotoxicity of 3-mercaptopropionic acid-modified quantum dots on Medicago sativa cells and tissues.
Methods in molecular biology (Clifton, N.J.) (2012-07-14)
Ana Raquel Santos, Ana Sofia Miguel, Pedro Fevereiro, Abel Oliva
ABSTRACT
Like most of the new technologies designed to interact with biological systems, the applications of -nanomaterials needs a proper assessment for their potential impacts. It is only through addressing the issues raised by toxicological studies that nanotechnology will be able to acquire its full potential. Here, we describe the detailed protocols to study the responses of plant cells to their exposure to nanoparticles, including viability, oxidative stress detection, and reactive oxygen species enzymatic detoxification, as well as particle uptake.
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%