Merck
  • CD109 overexpression ameliorates skin fibrosis in a mouse model of bleomycin-induced scleroderma.
CD109 overexpression ameliorates skin fibrosis in a mouse model of bleomycin-induced scleroderma.
Arthritis and rheumatism (2013-02-26)
Joshua Vorstenbosch, Hasan Al-Ajmi, Sebastian Winocour, Alissa Trzeciak, Lucie Lessard, Anie Philip
ABSTRACT
Transforming growth factor β (TGFβ) is a profibrotic cytokine, and its aberrant function is implicated in several types of fibrotic pathologies including scleroderma (systemic sclerosis [SSc]). Multiple lines of evidence show that increased TGFβ signaling contributes to progressive fibrosis in SSc by promoting fibroblast activation, excessive extracellular matrix (ECM) deposition, and dermal thickening. We have previously identified CD109 as a TGFβ coreceptor and have shown that it antagonizes TGFβ signaling and TGFβ-induced ECM expression in vitro in human keratinocytes and fibroblasts. The aim of the present study was to examine the ability of CD109 to prevent skin fibrosis in a mouse model of bleomycin-induced SSc. Transgenic mice overexpressing CD109 in the epidermis and their wild-type (WT) littermates were injected with bleomycin in phosphate buffered saline (PBS) or with PBS alone every other day for 21 days or 28 days. Dermal thickness and collagen deposition were determined histologically using Masson's trichrome and picrosirius red staining. In addition, collagen and fibronectin content was analyzed using Western blotting, and activation of TGFβ signaling was examined by determining phospho-Smad2 and phospho-Smad3 levels using Western blotting and immunohistochemistry. Transgenic mice overexpressing CD109 in the epidermis showed resistance to bleomycin-induced skin fibrosis, as evidenced by a significant decrease in dermal thickness, collagen crosslinking, collagen and fibronectin content, and phospho-Smad2/3 levels, as compared to their WT littermates. Our findings suggest that CD109 inhibits TGFβ signaling and fibrotic responses in experimental murine scleroderma. They also suggest that CD109 regulates dermal-epidermal interactions to decrease extracellular matrix synthesis in the dermis. Thus, CD109 is a potential molecular target for therapeutic intervention in scleroderma.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, suitable for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture