Merck
  • Description and preliminary evaluation of a new ultrasonic atomizer for spray-congealing processes.
Description and preliminary evaluation of a new ultrasonic atomizer for spray-congealing processes.
International journal of pharmaceutics (1999-06-11)
L Rodriguez, N Passerini, C Cavallari, M Cini, P Sancin, A Fini
ABSTRACT
A new atomizer that operates with ultrasonic energy is described. This apparatus is intended to obtain microparticulate drug delivery systems through spray-congealing or spray-drying technologies. In this work, some experimental results are reported on model systems submitted to spray-congealing. The formulations under examination contained theophylline and fenbufen as model drugs and stearic acid, carnauba wax, Cutina HR(R) and Compritol 888 ATO(R) as low melting excipients. Non-aggregate and spherical-shaped microparticles were obtained with all the materials tested; moreover, they had smooth surface and good flowability. The particle sizes depend on the amount of drug present and in each case the maximum size value of the distribution frequency was found to be 375 mu. In vitro release of the drug depends on its solubility and on the excipient lipophilicity. The results suggest that the ultrasound-assisted atomizer could be proposed as a possible alternative to traditional atomizers used for spray-congealing in the pharmaceutical field.