Merck
  • Nitric oxide donors enhance calcitonin gene-related peptide-induced elevations of cyclic AMP in vascular smooth muscle cells.
Nitric oxide donors enhance calcitonin gene-related peptide-induced elevations of cyclic AMP in vascular smooth muscle cells.
European journal of pharmacology (1999-08-17)
L F Lu, R R Fiscus
ABSTRACT
Vasorelaxant effects of calcitonin gene-related peptide (CGRP) are dependent on endothelium-derived nitric oxide (NO) in some arteries. The mechanism involved is still not clear. In the present study, we used NO donors (sodium nitroprusside (SNP) and 6-(2-hydroxy-1-methyl-2-nitrisohydrazino)-N-methyl-1-hyxanamine (NOC-9)), cyclic GMP elevator (brain natriuretic peptide (BNP)) and a selective type III (cyclic GMP-inhibited) phosphodiesterase (PDE) inhibitor 5-(4-acetamidophenyl)pyrazin-2(1H)-one (SK&F94120) to investigate involvement of NO, cyclic GMP and type III PDE in CGRP-induced accumulation of cyclic AMP in cultured rat aortic smooth muscle cells. SNP (10 microM), NOC-9 (10 microM) and BNP (1 microM) all increased intracellular cyclic GMP to similar levels (2- to 2.5-fold above basal) and caused significant enhancement of CGRP (10 nM)-induced cyclic AMP accumulation similar to that caused by 10 microM SK&F 94120. The data are therefore consistent with our hypothesis that the mechanism of endothelium-dependent vasorelaxation effect of CGRP involves cyclic GMP-mediated inhibition of type III PDE and subsequent accumulation of cyclic AMP in smooth muscle cells.
Sigma-Aldrich
MAHMA NONOate