Merck
  • A method for genotyping murine arylamine N-acetyltransferase type 2 (NAT2): a gene expressed in preimplantation embryonic stem cells encoding an enzyme acetylating the folate catabolite p-aminobenzoylglutamate.
A method for genotyping murine arylamine N-acetyltransferase type 2 (NAT2): a gene expressed in preimplantation embryonic stem cells encoding an enzyme acetylating the folate catabolite p-aminobenzoylglutamate.
Biochemical pharmacology (1999-08-17)
M Payton, V Smelt, A Upton, E Sim
ABSTRACT
Mice have three arylamine N-acetyltransferase (NAT) isoenzymes (NAT1, NAT2, and NAT3) of which NAT2 is known to be polymorphic. Humans have two polymorphic isoenzymes, NAT1 and NAT2. The isoenzymes mouse NAT1 and human NAT2 are expressed predominantly in the liver and intestine and are involved in drug and xenobiotic metabolism. Mouse NAT2 and human NAT1 have a widespread tissue distribution and the folate catabolite p-aminobenzoylglutamate (pAB-Glu) has been proposed as a candidate endogenous substrate. All mice have detectable NAT2 activity, although inbred mouse strains have either a fast or slow acetylator phenotype conferred by the presence of either NAT2*8 (fast) or NAT2*9 (slow) alleles at the NAT2 locus. In this report, we describe a simple method for distinguishing these murine alleles by polymerase chain reaction followed by restriction fragment length polymorphism analysis. We compared the tissue distribution of the acetylation activity found in both fast (C57BL/6J) and slow (A/J) acetylating strains of mice using pAB-Glu and p-aminobenzoic acid as probe substrates. It has previously been demonstrated that murine NAT2 is expressed in the neural tube prior to closure (Stanley L, Copp A, Rolls S, Smelt V, Perry VH and Sim E, Teratology 58: 174-182, 1998). We demonstrate here that murine NAT2 is expressed in preimplantation embryonic stem cells. Murine NAT2 is likely to be expressed prior to neurulation and this may be important in view of the protective role of folate in neural tube development.
Product Description
Folic acid impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-(4-Aminobenzoyl)-L-glutamic acid, ≥98% (TLC)