Merck
Characterization of T cell responses to fragrances.
Toxicology and applied pharmacology (2001-04-21)
S Sieben, M Hertl, T Al Masaoudi, H F Merk, B Blömeke
ABSTRACT
Fragrances are worldwide a major cause of allergic contact dermatitis (ACD), a delayed-type hypersensitivity reaction mediated by T lymphocytes. We investigated T cell responses to fragrances using peripheral blood mononuclear cells (PBMC) and T cells from skin lesions of fragrance-allergic patients. The components of a fragrance mixture (eugenol, isoeugenol, geraniol, oak moss, alpha-amyl cinnamic aldehyde, cinnamic aldehyde, cinnamic alcohol, and hydroxycitronellal) that is commonly used in the patch test were studied in vitro in the lymphocyte transformation test (LTT). PBMC from fragrance-allergic patients (n = 32) showed significant stimulations to all eight fragrances. The calculated stimulation indices (SI) varied between 2.1 and 21.8. The influence of metabolic enzymes on T cell stimulation was studied for two fragrances. Interestingly, stimulation of eugenol and isoeugenol was increased in the presence of antigen-modified human liver microsomes (CYP450) or recombinant CYP1A1 in five of seven cases. Furthermore, we established 18 T cell clones (TCC) from a skin lesion reacting specifically to eugenol. FACS analysis revealed that the majority (n = 15, 83%) of TCC were CD3(+), CD4(+), and HLA-DR(+). Seventeen percent (n = 3) of the clones were CD8(+). TCC (n = 4) released significant amounts of IL-2 and IFN-gamma but no IL-4 and IL-5. In addition, CD4(+) TCC (n = 3) showed antigen-induced cytotoxic activities against autologous B cells. In summary, we demonstrated for the first time that fragrance-specific CD4(+) and CD8(+) T lymphocytes are present in fragrance-allergic individuals. In addition, our results suggest that CYPs can be involved in the formation of the nominative antigen.
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG
Sigma-Aldrich
7-Hydroxycitronellal, Odorant used in allergy studies